Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CELF1
Entrez Gene ID
10658
Gene Name
CUGBP, Elav-like family member 1
Gene Aliases
BRUNOL2, CUG-BP, CUGBP, CUGBP1, EDEN-BP, NAB50, NAPOR, hNab50
Chromosome Location
Chr.11: 47465937 - 47565569 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001025596.2 NP_001020767.1 9 - 10 1012 58
NM_001172639.1 NP_001166110.1 12 - 13 1974 58
NM_001172640.1 NP_001166111.1 9 - 10 1009 58
NM_006560.3 NP_006551.1 10 - 11 1249 58
NM_198700.2 NP_941989.1 10 - 11 1194 58
XM_011519848.2 XP_011518150.1 10 - 11 1087 58
XM_011519850.2 XP_011518152.2 10 - 11 1323 58
XM_011519852.2 XP_011518154.1 12 - 13 1435 58
XM_011519854.1 XP_011518156.1 14 - 15 1543 58
XM_011519855.1 XP_011518157.1 10 - 11 1264 58
XM_011519856.1 XP_011518158.1 11 - 12 1345 58
XM_011519857.1 XP_011518159.1 10 - 11 1216 58
XM_011519858.1 XP_011518160.1 10 - 11 1311 58
XM_011519859.1 XP_011518161.1 14 - 15 1531 58
XM_017017101.1 XP_016872590.1 13 - 14 1427 58
XM_017017102.1 XP_016872591.1 12 - 13 1346 58
XM_017017103.1 XP_016872592.1 11 - 12 1239 58
XM_017017104.1 XP_016872593.1 11 - 12 1239 58
XM_017017105.1 XP_016872594.1 11 - 12 1236 58
XM_017017106.1 XP_016872595.1 13 - 14 1424 58
XM_017017107.1 XP_016872596.1 11 - 12 1236 58
XM_017017108.1 XP_016872597.1 11 - 12 1227 58
XM_017017109.1 XP_016872598.1 10 - 11 1314 58
XM_017017110.1 XP_016872599.1 11 - 12 1227 58
XM_017017111.1 XP_016872600.1 11 - 12 1224 58
XM_017017112.1 XP_016872601.1 11 - 12 1224 58
XM_017017113.1 XP_016872602.1 11 - 12 1224 58
XM_017017114.1 XP_016872603.1 11 - 12 1224 58
XM_017017115.1 XP_016872604.1 12 - 13 1267 58
XM_017017116.1 XP_016872605.1 11 - 12 1302 58
XM_017017117.1 XP_016872606.1 10 - 11 1063 58
XM_017017118.1 XP_016872607.1 10 - 11 1185 58
XM_017017119.1 XP_016872608.1 12 - 13 1404 58
XM_017017120.1 XP_016872609.1 10 - 11 1216 58
XM_017017121.1 XP_016872610.1 10 - 11 1311 58
XM_017017122.1 XP_016872611.1 10 - 11 1216 58
XM_017017123.1 XP_016872612.1 10 - 11 1264 58
XM_017017124.1 XP_016872613.1 12 - 13 1355 58
XM_017017125.1 XP_016872614.1 10 - 11 1084 58
XM_017017126.1 XP_016872615.1 10 - 11 1213 58
XM_017017127.1 XP_016872616.1 10 - 11 1237 58
XM_017017128.1 XP_016872617.1 10 - 11 1261 58
XM_017017129.1 XP_016872618.1 11 - 12 1259 58
XM_017017130.1 XP_016872619.1 10 - 11 1213 58
XM_017017131.1 XP_016872620.1 10 - 11 1237 58
XM_017017132.1 XP_016872621.1 10 - 11 1261 58
XM_017017133.1 XP_016872622.1 10 - 11 1043 58
XM_017017134.1 XP_016872623.1 11 - 12 1182 58
XM_017017135.1 XP_016872624.1 10 - 11 1201 58
GenBank mRNA AB210019.1 - 12 - 13 1974 58
AF248648.1 - 9 - 10 1020 58
AF267533.1 - 9 - 10 1013 58
AF267534.1 - 9 - 10 1004 58
AJ007988.1 - 9 - 10 1002 58
AK304430.1 - 9 - 10 952 58
AK307679.1 - 4 - 5 474 58
BC031079.1 - 10 - 11 1194 58
U63289.1 - 10 - 11 1127 58

View Full Product Details