Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CELF1
Entrez Gene ID
10658
Gene Name
CUGBP, Elav-like family member 1
Gene Aliases
BRUNOL2, CUG-BP, CUGBP, CUGBP1, EDEN-BP, NAB50, NAPOR, hNab50
Chromosome Location
Chr.11: 47465937 - 47565569 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001025596.2 NP_001020767.1 2 - 3 276 76
NM_001172639.1 NP_001166110.1 5 - 6 1241 76
NM_001172640.1 NP_001166111.1 2 - 3 276 76
NM_006560.3 NP_006551.1 3 - 4 525 76
NM_198700.2 NP_941989.1 3 - 4 467 76
XM_011519848.2 XP_011518150.1 3 - 4 348 76
XM_011519850.2 XP_011518152.2 3 - 4 587 76
XM_011519852.2 XP_011518154.1 5 - 6 696 76
XM_011519854.1 XP_011518156.1 7 - 8 804 76
XM_011519855.1 XP_011518157.1 3 - 4 525 76
XM_011519856.1 XP_011518158.1 4 - 5 606 76
XM_011519857.1 XP_011518159.1 3 - 4 477 76
XM_011519858.1 XP_011518160.1 3 - 4 572 76
XM_011519859.1 XP_011518161.1 7 - 8 804 76
XM_017017101.1 XP_016872590.1 6 - 7 688 76
XM_017017102.1 XP_016872591.1 5 - 6 607 76
XM_017017103.1 XP_016872592.1 4 - 5 500 76
XM_017017104.1 XP_016872593.1 4 - 5 500 76
XM_017017105.1 XP_016872594.1 4 - 5 500 76
XM_017017106.1 XP_016872595.1 6 - 7 688 76
XM_017017107.1 XP_016872596.1 4 - 5 500 76
XM_017017108.1 XP_016872597.1 4 - 5 500 76
XM_017017109.1 XP_016872598.1 3 - 4 587 76
XM_017017110.1 XP_016872599.1 4 - 5 500 76
XM_017017111.1 XP_016872600.1 4 - 5 500 76
XM_017017112.1 XP_016872601.1 4 - 5 500 76
XM_017017113.1 XP_016872602.1 4 - 5 500 76
XM_017017114.1 XP_016872603.1 4 - 5 500 76
XM_017017115.1 XP_016872604.1 5 - 6 528 76
XM_017017116.1 XP_016872605.1 4 - 5 563 76
XM_017017117.1 XP_016872606.1 3 - 4 324 76
XM_017017118.1 XP_016872607.1 3 - 4 446 76
XM_017017119.1 XP_016872608.1 5 - 6 665 76
XM_017017120.1 XP_016872609.1 3 - 4 477 76
XM_017017121.1 XP_016872610.1 3 - 4 572 76
XM_017017122.1 XP_016872611.1 3 - 4 477 76
XM_017017123.1 XP_016872612.1 3 - 4 525 76
XM_017017124.1 XP_016872613.1 5 - 6 619 76
XM_017017125.1 XP_016872614.1 3 - 4 348 76
XM_017017126.1 XP_016872615.1 3 - 4 477 76
XM_017017127.1 XP_016872616.1 3 - 4 501 76
XM_017017128.1 XP_016872617.1 3 - 4 525 76
XM_017017129.1 XP_016872618.1 4 - 5 523 76
XM_017017130.1 XP_016872619.1 3 - 4 477 76
XM_017017131.1 XP_016872620.1 3 - 4 501 76
XM_017017132.1 XP_016872621.1 3 - 4 525 76
XM_017017133.1 XP_016872622.1 3 - 4 316 76
XM_017017134.1 XP_016872623.1 4 - 5 455 76
XM_017017135.1 XP_016872624.1 3 - 4 477 76
GenBank mRNA AB210019.1 - 5 - 6 1241 76
AF248648.1 - 2 - 3 284 76
AF267533.1 - 2 - 3 277 76
AF267534.1 - 2 - 3 277 76
AJ007988.1 - 2 - 3 266 76
AK304430.1 - 2 - 3 219 76
BC031079.1 - 3 - 4 467 76
CB960956.1 - 4 - 5 428 76
DA320861.1 - 3 - 4 526 76
DB162286.1 - 3 - 4 425 76
U63289.1 - 3 - 4 403 76

View Full Product Details