Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MECOM
Entrez Gene ID
2122
Gene Name
MDS1 and EVI1 complex locus
Gene Aliases
AML1-EVI-1, EVI1, MDS1, MDS1-EVI1, PRDM3, RUSAT2
Chromosome Location
Chr.3: 169083499 - 169663781 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_004991.3 NP_004982.2 1 - 2 440 61
XM_005247213.3 XP_005247270.1 1 - 2 446 61
XM_005247214.3 XP_005247271.1 1 - 2 446 61
XM_005247215.3 XP_005247272.1 1 - 2 445 61
XM_005247224.3 XP_005247281.1 1 - 2 446 61
XM_005247225.3 XP_005247282.1 1 - 2 445 61
XM_005247226.3 XP_005247283.1 1 - 2 323 61
XM_017005877.1 XP_016861366.1 1 - 2 445 61
GenBank mRNA AK304098.1 - 1 - 2 386 61
AK308404.1 - 1 - 2 85 61
AL700380.1 - 1 - 2 380 61
BC069498.1 - 1 - 2 289 61
CR541866.1 - 1 - 2 37 61
CR541886.1 - 1 - 2 37 61
DC347146.1 - 1 - 2 435 61
DC348381.1 - 1 - 2 426 61
U43292.1 - 1 - 2 344 61

View Full Product Details