The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001184906.1 | NP_001171835.1 | 7 - 8 | 783 | 104 |
NM_032875.2 | NP_116264.2 | 8 - 9 | 879 | 104 | |
XM_005257746.3 | XP_005257803.2 | 8 - 9 | 1070 | 104 | |
XM_005257747.3 | XP_005257804.2 | 7 - 8 | 851 | 104 | |
GenBank mRNA | AK123048.1 | - | 8 - 9 | 880 | 104 |
AK291844.1 | - | 8 - 9 | 890 | 104 | |
AK312612.1 | - | 8 - 9 | 860 | 104 | |
BC007557.1 | - | 8 - 9 | 846 | 104 | |
DQ223959.1 | - | 7 - 8 | 750 | 104 |