Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CES4A
Entrez Gene ID
283848
Gene Name
carboxylesterase 4A
Gene Aliases
CES6, CES8
Chromosome Location
Chr.16: 66988589 - 67009758 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_173815.6 NP_776176.5 2 - 3 442 84
XM_005255896.4 XP_005255953.1 2 - 3 439 84
XM_011523026.2 XP_011521328.1 2 - 3 439 84
XM_011523028.2 XP_011521330.1 2 - 3 447 84
XM_011523030.2 XP_011521332.1 2 - 3 439 84
GenBank mRNA AK296064.1 - 2 - 3 362 84
BC084555.1 - 1 - 2 147 84
DC339496.1 - 2 - 3 442 84

View Full Product Details