The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001190201.1 | NP_001177130.1 | 2 - 3 | 407 | 90 |
NM_173815.6 | NP_776176.5 | 4 - 5 | 726 | 90 | |
XM_005255896.4 | XP_005255953.1 | 4 - 5 | 723 | 90 | |
XM_011523023.2 | XP_011521325.1 | 5 - 6 | 843 | 90 | |
XM_011523030.2 | XP_011521332.1 | 4 - 5 | 723 | 90 | |
XM_017023160.1 | XP_016878649.1 | 5 - 6 | 843 | 90 | |
XM_017023161.1 | XP_016878650.1 | 5 - 6 | 843 | 90 | |
XM_017023164.1 | XP_016878653.1 | 5 - 6 | 843 | 90 | |
XM_017023165.1 | XP_016878654.1 | 5 - 6 | 843 | 90 | |
XM_017023167.1 | XP_016878656.1 | 2 - 3 | 807 | 90 | |
XM_017023168.1 | XP_016878657.1 | 2 - 3 | 813 | 90 | |
GenBank mRNA | AK293061.1 | - | 2 - 3 | 255 | 90 |
AK296064.1 | - | 4 - 5 | 646 | 90 | |
AK300792.1 | - | 2 - 3 | 367 | 90 | |
BC064573.1 | - | 2 - 3 | 187 | 90 | |
BC084555.1 | - | 3 - 4 | 431 | 90 | |
DC306014.1 | - | 2 - 3 | 407 | 90 |