The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015512.4 | NP_056327.4 | 18 - 19 | 3279 | 60 |
XM_017006129.1 | XP_016861618.1 | 19 - 20 | 6167 | 60 | |
XM_017006130.1 | XP_016861619.1 | 19 - 20 | 6166 | 60 | |
XM_017006131.1 | XP_016861620.1 | 19 - 20 | 6167 | 60 | |
XM_017006132.1 | XP_016861621.1 | 19 - 20 | 6168 | 60 | |
XM_017006133.1 | XP_016861622.1 | 19 - 20 | 6168 | 60 | |
GenBank mRNA | AB037831.3 | - | 18 - 19 | 3279 | 60 |
AB290163.1 | - | 18 - 19 | 3279 | 60 | |
AK125990.1 | - | 19 - 20 | 3183 | 60 |