The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015512.4 | NP_056327.4 | 61 - 62 | 10004 | 70 |
XM_017006129.1 | XP_016861618.1 | 63 - 64 | 12961 | 70 | |
XM_017006130.1 | XP_016861619.1 | 62 - 63 | 12891 | 70 | |
XM_017006131.1 | XP_016861620.1 | 62 - 63 | 12835 | 70 | |
XM_017006132.1 | XP_016861621.1 | 63 - 64 | 12962 | 70 | |
GenBank mRNA | AB037831.3 | - | 61 - 62 | 10004 | 70 |
AB290163.1 | - | 60 - 61 | 10199 | 70 | |
AK074130.2 | - | 8 - 9 | 1529 | 70 | |
AY221994.1 | - | 17 - 18 | 3047 | 70 |