The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001077693.3 | NP_001071161.1 | 5 - 6 | 336 | 78 |
NM_001293739.1 | NP_001280668.1 | 5 - 6 | 336 | 78 | |
NR_121659.1 | 5 - 6 | 336 | 78 | ||
GenBank mRNA | BC146912.1 | - | 5 - 6 | 308 | 78 |
BC146913.1 | - | 5 - 6 | 308 | 78 | |
BF526332.1 | - | - | 356 | 79 | |
BI834795.1 | - | 5 - 6 | 286 | 78 | |
DQ462572.1 | - | 5 - 6 | 330 | 78 | |
EU025066.1 | - | 5 - 6 | 336 | 78 |