The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003265.2 | NP_003256.1 | 2 - 3 | 536 | 135 |
GenBank mRNA | AB445631.1 | - | 1 - 2 | 435 | 135 |
AK314208.1 | - | 2 - 3 | 535 | 135 | |
BC059372.1 | - | 2 - 3 | 535 | 135 | |
BC094737.1 | - | 2 - 3 | 535 | 135 | |
BC096333.1 | - | 2 - 3 | 513 | 135 | |
BC096334.3 | - | 2 - 3 | 513 | 135 | |
BC096335.1 | - | 2 - 3 | 513 | 135 | |
DA374150.1 | - | 2 - 3 | 536 | 135 | |
DQ445682.1 | - | 2 - 3 | 506 | 135 | |
U88879.1 | - | 2 - 3 | 508 | 135 |