Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- LRRC74B
- Entrez Gene ID
-
400891
- Gene Name
- leucine rich repeat containing 74B
- Gene Aliases
-
-
- Chromosome Location
-
Chr.22: 21045960 - 21064168 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_027006.1 |
|
6 - 7 |
1955 |
147 |
GenBank mRNA |
AK128646.1 |
- |
6 - 7 |
1934 |
147 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
purinergic receptor P2X 6 pseudogene |
P2RX6P |
|
-9.1kb
|
Anti-sense |
Chr. - 21042392 - 21044249 on Build GRCh38 |
Downstream |
breakpoint cluster region pseudogene 2 |
BCRP2 |
BCR-2, BCR2, BCRL2 |
49.0kb
|
Sense |
Chr. - 21103016 - 21122286 on Build GRCh38 |
View Full Product Details