Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- PXK
- Entrez Gene ID
-
54899
- Gene Name
- PX domain containing serine/threonine kinase like
- Gene Aliases
-
MONaKA
- Chromosome Location
-
Chr.3: 58332868 - 58426127 on Build GRCh38
View Full Product Details