The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001286762.1 | NP_001273691.1 | 11 - 12 | 2116 | 105 |
GenBank mRNA | AK025215.1 | - | 11 - 12 | 1865 | 105 |