Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
NDUFAF3
Entrez Gene ID
25915
Gene Name
NADH:ubiquinone oxidoreductase complex assembly factor 3
Gene Aliases
2P1, C3orf60, E3-3
Chromosome Location
Chr.3: 49020475 - 49023495 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_199069.1 NP_951032.1 3 - 4 842 108
NM_199070.1 NP_951033.1 3 - 4 405 108
NM_199073.1 NP_951047.1 3 - 4 476 108
NM_199074.1 NP_951056.1 3 - 4 357 108
GenBank mRNA AL049955.1 - 3 - 4 409 108
BC002873.2 - 3 - 4 392 108
BC111004.1 - 3 - 4 1472 108
BM354128.1 - 2 - 3 448 108
BM985326.1 - 2 - 3 108
BQ652614.1 - 3 - 4 357 108
CD358942.1 - 3 - 4 546 108

View Full Product Details