Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
BPI
Entrez Gene ID
671
Gene Name
bactericidal/permeability-increasing protein
Gene Aliases
BPIFD1, rBPI
Chromosome Location
Chr.20: 38304150 - 38337503 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001725.2 NP_001716.2 7 - 8 824 62
GenBank mRNA AF322588.1 - 7 - 8 750 62
AK296568.1 - 7 - 8 669 62
AK315328.1 - 7 - 8 791 62
AX713278.1 - 7 - 8 792 62
AX746123.1 - 7 - 8 792 62
AX752831.1 - 7 - 8 792 62
BC040955.1 - 7 - 8 799 62
DQ414688.1 - 7 - 8 762 62
J04739.1 - 7 - 8 792 62

View Full Product Details