The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001318253.1 | NP_001305182.1 | 17 - 18 | 2275 | 85 |
NM_133171.4 | NP_573403.1 | 17 - 18 | 2207 | 85 | |
NM_182764.2 | NP_877496.1 | 16 - 17 | 2103 | 85 | |
XM_005260496.3 | XP_005260553.1 | 17 - 18 | 1772 | 85 | |
XM_005260498.3 | XP_005260555.1 | 16 - 17 | 2134 | 85 | |
XM_005260499.3 | XP_005260556.1 | 16 - 17 | 2105 | 85 | |
XM_005260500.3 | XP_005260557.1 | 15 - 16 | 1633 | 85 | |
XM_005260501.4 | XP_005260558.1 | 16 - 17 | 1643 | 85 | |
XM_006723854.3 | XP_006723917.1 | 16 - 17 | 1706 | 85 | |
XM_017028009.1 | XP_016883498.1 | 16 - 17 | 2078 | 85 | |
XM_017028010.1 | XP_016883499.1 | 15 - 16 | 1581 | 85 | |
XM_017028011.1 | XP_016883500.1 | 15 - 16 | 2034 | 85 | |
GenBank mRNA | AB058737.1 | - | 16 - 17 | 1718 | 85 |
AF398886.1 | - | 15 - 16 | 1646 | 85 | |
AF417861.1 | - | 15 - 16 | 1578 | 85 | |
AK021718.1 | - | 6 - 7 | 774 | 85 | |
AK022731.1 | - | 4 - 5 | 489 | 85 | |
AK023103.1 | - | 5 - 6 | 534 | 85 | |
AK293861.1 | - | 15 - 16 | 1671 | 85 | |
AK299318.1 | - | 19 - 20 | 1818 | 85 | |
AK302710.1 | - | 7 - 8 | 1010 | 85 | |
AL713765.1 | - | 6 - 7 | 752 | 85 | |
BC000143.4 | - | 16 - 17 | 1783 | 85 | |
BC021079.2 | - | 6 - 7 | 770 | 85 | |
BX648819.1 | - | 16 - 17 | 1684 | 85 |