The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001318253.1 | NP_001305182.1 | 19 - 20 | 2500 | 91 |
NM_133171.4 | NP_573403.1 | 19 - 20 | 2432 | 91 | |
NM_182764.2 | NP_877496.1 | 18 - 19 | 2328 | 91 | |
XM_005260496.3 | XP_005260553.1 | 19 - 20 | 1997 | 91 | |
XM_005260498.3 | XP_005260555.1 | 18 - 19 | 2359 | 91 | |
XM_005260499.3 | XP_005260556.1 | 18 - 19 | 2330 | 91 | |
XM_005260500.3 | XP_005260557.1 | 17 - 18 | 1858 | 91 | |
XM_005260501.4 | XP_005260558.1 | 18 - 19 | 1868 | 91 | |
XM_006723854.3 | XP_006723917.1 | 18 - 19 | 1931 | 91 | |
XM_017028009.1 | XP_016883498.1 | 18 - 19 | 2303 | 91 | |
XM_017028010.1 | XP_016883499.1 | 17 - 18 | 1806 | 91 | |
XM_017028011.1 | XP_016883500.1 | 17 - 18 | 2259 | 91 | |
GenBank mRNA | AB058737.1 | - | 18 - 19 | 1943 | 91 |
AF398886.1 | - | 17 - 18 | 1871 | 91 | |
AF417861.1 | - | 17 - 18 | 1803 | 91 | |
AK021718.1 | - | 8 - 9 | 999 | 91 | |
AK023103.1 | - | 7 - 8 | 759 | 91 | |
AK293861.1 | - | 17 - 18 | 1896 | 91 | |
AK299318.1 | - | 21 - 22 | 2043 | 91 | |
AK302710.1 | - | 9 - 10 | 1235 | 91 | |
AL713765.1 | - | 8 - 9 | 977 | 91 | |
BC000143.4 | - | 18 - 19 | 2008 | 91 | |
BC021079.2 | - | 8 - 9 | 995 | 91 | |
BX648819.1 | - | 18 - 19 | 1909 | 91 |