The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_133171.4 | NP_573403.1 | 4 - 5 | 747 | 69 |
NM_182764.2 | NP_877496.1 | 3 - 4 | 643 | 69 | |
XM_005260496.3 | XP_005260553.1 | 4 - 5 | 312 | 69 | |
XM_005260501.4 | XP_005260558.1 | 4 - 5 | 283 | 69 | |
XM_006723854.3 | XP_006723917.1 | 3 - 4 | 246 | 69 | |
XM_017028009.1 | XP_016883498.1 | 4 - 5 | 718 | 69 | |
XM_017028010.1 | XP_016883499.1 | 3 - 4 | 221 | 69 | |
GenBank mRNA | AB058737.1 | - | 3 - 4 | 258 | 69 |
AF417861.1 | - | 2 - 3 | 118 | 69 | |
AK057032.1 | - | 4 - 5 | 271 | 69 | |
AK293861.1 | - | 3 - 4 | 311 | 69 | |
AK299318.1 | - | 5 - 6 | 322 | 69 | |
BX648819.1 | - | 3 - 4 | 224 | 69 |