Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
POU2F1
Entrez Gene ID
5451
Gene Name
POU class 2 homeobox 1
Gene Aliases
OCT1, OTF1, oct-1B
Chromosome Location
Chr.1: 167220829 - 167427345 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001198783.1 NP_001185712.1 9 - 10 1186 67
NM_001198786.1 NP_001185715.1 8 - 9 864 67
NM_002697.3 NP_002688.3 9 - 10 1053 67
NR_037163.1 10 - 11 1097 67
XM_011509653.2 XP_011507955.1 10 - 11 1005 67
XM_011509654.2 XP_011507956.1 9 - 10 966 67
XM_011509655.1 XP_011507957.1 10 - 11 1005 67
XM_017001508.1 XP_016856997.1 10 - 11 1005 67
GenBank mRNA AK091438.1 - 11 - 12 1481 67
AK302525.1 - 11 - 12 1159 67
AK303201.1 - 8 - 9 802 67
AK308004.1 - 10 - 11 1092 67
AY113189.1 - 9 - 10 1005 67
BC001664.2 - 9 - 10 969 67
BC003571.1 - 9 - 10 969 67
BC007388.2 - 5 - 6 639 67
BC052274.1 - 10 - 11 1077 67
KT438684.1 - 11 - 12 1164 67
X13403.1 - 9 - 10 974 67

View Full Product Details