Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
POU2F1
Entrez Gene ID
5451
Gene Name
POU class 2 homeobox 1
Gene Aliases
OCT1, OTF1, oct-1B
Chromosome Location
Chr.1: 167220829 - 167427345 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001198783.1 NP_001185712.1 15 - 16 2190 103
NM_001198786.1 NP_001185715.1 14 - 15 1868 103
NM_002697.3 NP_002688.3 15 - 16 2057 103
NR_037163.1 16 - 17 2101 103
XM_011509653.2 XP_011507955.1 16 - 17 2009 103
XM_011509654.2 XP_011507956.1 15 - 16 1970 103
GenBank mRNA AK091438.1 - 17 - 18 2485 103
AK303201.1 - 14 - 15 1806 103
AY113189.1 - 15 - 16 2009 103
BC001664.2 - 15 - 16 1973 103
BC003571.1 - 15 - 16 1973 103
BC052274.1 - 16 - 17 2081 103
KT438684.1 - 17 - 18 2168 103
X13403.1 - 15 - 16 1978 103

View Full Product Details