The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_021147.4 | NP_066970.3 | 1 - 2 | 579 | 78 |
GenBank mRNA | AK026075.1 | - | 1 - 2 | 542 | 78 |