The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_017415.2 | NP_059111.2 | 1 - 2 | 461 | 66 |
GenBank mRNA | AB032955.1 | - | 1 - 2 | 231 | 66 |
AF208068.1 | - | 1 - 2 | 145 | 66 | |
AK314707.1 | - | 1 - 2 | 202 | 66 | |
BC034035.1 | - | 1 - 2 | 460 | 66 | |
DA083147.1 | - | 1 - 2 | 461 | 66 |