The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_133443.3 | NP_597700.1 | 1 - 2 | 85 | 66 |
GenBank mRNA | AY029173.2 | - | 1 - 2 | 69 | 66 |
HY117879.1 | - | 1 - 2 | 85 | 66 |