The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_033226.2 | NP_150229.2 | 13 - 14 | 2333 | 137 |
GenBank mRNA | AF395908.1 | - | 13 - 14 | 1993 | 137 |
AF395909.1 | - | 13 - 14 | 1993 | 137 | |
AF411577.1 | - | 13 - 14 | 1993 | 137 | |
AF411578.1 | - | 13 - 14 | 1993 | 137 | |
AY040220.1 | - | 13 - 14 | 2333 | 137 | |
AY196326.1 | - | 12 - 13 | 2039 | 137 | |
BC036378.1 | - | 14 - 15 | 2418 | 137 |