The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_033226.2 | NP_150229.2 | 24 - 25 | 3818 | 63 |
GenBank mRNA | AF395908.1 | - | 24 - 25 | 3499 | 63 |
AF411577.1 | - | 24 - 25 | 3499 | 63 | |
AF411578.1 | - | 24 - 25 | 3499 | 63 | |
AK307109.1 | - | 2 - 3 | 289 | 63 | |
AK308599.1 | - | 2 - 3 | 306 | 63 | |
AY040220.1 | - | 24 - 25 | 3818 | 63 | |
AY196326.1 | - | 22 - 23 | 3482 | 63 | |
BC036378.1 | - | 25 - 26 | 3924 | 63 | |
DA096073.1 | - | 2 - 3 | 306 | 63 |