Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ZCWPW1
Entrez Gene ID
55063
Gene Name
zinc finger CW-type and PWWP domain containing 1
Gene Aliases
ZCW1
Chromosome Location
Chr.7: 100400826 - 100429239 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001258008.1 NP_001244937.1 12 - 13 1553 91
NM_017984.4 NP_060454.3 12 - 13 1550 91
XM_005250479.2 XP_005250536.1 12 - 13 1987 91
XM_006716035.3 XP_006716098.1 12 - 13 1987 91
XM_006716036.2 XP_006716099.1 12 - 13 1491 91
XM_006716037.3 XP_006716100.1 12 - 13 1984 91
XM_006716038.3 XP_006716101.1 12 - 13 1984 91
GenBank mRNA AK000919.1 - 7 - 8 937 91
AK093038.1 - 12 - 13 1491 91
AK300745.1 - 12 - 13 1409 91
AK302108.1 - 11 - 12 1287 91
AL136735.1 - 12 - 13 1421 91
BC002725.1 - 9 - 10 1251 91
KU178677.1 - 7 - 8 813 91
KU178678.1 - 7 - 8 816 91
KU178679.1 - 7 - 8 813 91
KU178680.1 - 7 - 8 816 91
KU178681.1 - 7 - 8 813 91

View Full Product Details