Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ZCWPW1
Entrez Gene ID
55063
Gene Name
zinc finger CW-type and PWWP domain containing 1
Gene Aliases
ZCW1
Chromosome Location
Chr.7: 100400826 - 100429239 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001258008.1 NP_001244937.1 1 - 2 241 115
NM_017984.4 NP_060454.3 1 - 2 241 115
XM_005250479.2 XP_005250536.1 1 - 2 672 115
XM_005250480.2 XP_005250537.1 1 - 2 672 115
XM_006716035.3 XP_006716098.1 1 - 2 672 115
XM_006716037.3 XP_006716100.1 1 - 2 672 115
XM_006716038.3 XP_006716101.1 1 - 2 672 115
XM_006716040.3 XP_006716103.1 1 - 2 672 115
XM_017012379.1 XP_016867868.1 1 - 2 672 115
GenBank mRNA AK299989.1 - 1 - 2 129 115
AK300745.1 - 1 - 2 97 115
AK302108.1 - 1 - 2 95 115
AK316553.1 - 1 - 2 429 115
AL136735.1 - 1 - 2 112 115
DB447154.1 - 1 - 2 241 115
DC413605.1 - 1 - 2 226 116

View Full Product Details