Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SEZ6L
Entrez Gene ID
23544
Gene Name
seizure related 6 homolog like
Gene Aliases
-
Chromosome Location
Chr.22: 26169474 - 26383597 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_021115.4 NP_066938.2 14 - 15 3030 81
XM_005261440.3 XP_005261497.1 13 - 14 2815 81
GenBank mRNA AB041736.1 - 14 - 15 2834 81
BC126115.1 - 14 - 15 2929 81
BM875686.1 - 4 - 5 309 81

View Full Product Details