The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001184773.1 | NP_001171702.1 | 6 - 7 | 1711 | 53 |
NM_001184774.1 | NP_001171703.1 | 6 - 7 | 1711 | 53 | |
NM_001184775.1 | NP_001171704.1 | 6 - 7 | 1711 | 53 | |
NM_001184776.1 | NP_001171705.1 | 6 - 7 | 1711 | 53 | |
NM_001184777.1 | NP_001171706.1 | 6 - 7 | 1711 | 53 | |
NM_021115.4 | NP_066938.2 | 6 - 7 | 1711 | 53 | |
XM_005261439.3 | XP_005261496.1 | 6 - 7 | 1687 | 53 | |
XM_005261440.3 | XP_005261497.1 | 6 - 7 | 1688 | 53 | |
XM_006724195.3 | XP_006724258.1 | 6 - 7 | 1687 | 53 | |
XM_011530037.2 | XP_011528339.1 | 6 - 7 | 1687 | 53 | |
XM_011530038.2 | XP_011528340.1 | 6 - 7 | 1687 | 53 | |
XM_011530039.2 | XP_011528341.1 | 6 - 7 | 1687 | 53 | |
XM_017028699.1 | XP_016884188.1 | 6 - 7 | 1688 | 53 | |
XM_017028700.1 | XP_016884189.1 | 6 - 7 | 1688 | 53 | |
GenBank mRNA | AB023144.2 | - | 6 - 7 | 1671 | 53 |
AB041736.1 | - | 6 - 7 | 1515 | 53 | |
AL035545.1 | - | 5 - 6 | 1576 | 53 | |
AL050253.1 | - | 5 - 6 | 1576 | 53 | |
AY358405.1 | - | 6 - 7 | 1711 | 53 | |
BC126115.1 | - | 6 - 7 | 1610 | 53 | |
BC143851.1 | - | 6 - 7 | 1610 | 53 | |
BC143854.1 | - | 6 - 7 | 1610 | 53 | |
CR456574.1 | - | 6 - 7 | 1527 | 53 |