Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SEZ6L
Entrez Gene ID
23544
Gene Name
seizure related 6 homolog like
Gene Aliases
-
Chromosome Location
Chr.22: 26169474 - 26383597 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001184773.1 NP_001171702.1 8 - 9 2074 72
NM_001184774.1 NP_001171703.1 8 - 9 2074 72
NM_001184775.1 NP_001171704.1 8 - 9 2074 72
NM_001184776.1 NP_001171705.1 8 - 9 2074 72
NM_001184777.1 NP_001171706.1 8 - 9 2074 72
NM_021115.4 NP_066938.2 8 - 9 2074 72
XM_005261439.3 XP_005261496.1 8 - 9 2050 72
XM_005261440.3 XP_005261497.1 8 - 9 2051 72
XM_006724195.3 XP_006724258.1 8 - 9 2050 72
XM_011530037.2 XP_011528339.1 8 - 9 2050 72
XM_011530038.2 XP_011528340.1 8 - 9 2050 72
XM_011530039.2 XP_011528341.1 8 - 9 2050 72
XM_017028699.1 XP_016884188.1 8 - 9 2051 72
XM_017028700.1 XP_016884189.1 8 - 9 2051 72
GenBank mRNA AB023144.2 - 8 - 9 2034 72
AB041736.1 - 8 - 9 1878 72
AL035545.1 - 7 - 8 1939 72
AL050253.1 - 7 - 8 1939 72
AY358405.1 - 8 - 9 2074 72
BC126115.1 - 8 - 9 1973 72
BC143851.1 - 8 - 9 1973 72
BC143854.1 - 8 - 9 1973 72
CR456574.1 - 8 - 9 1890 72

View Full Product Details