The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002726.4 | NP_002717.3 | 1 - 2 | 275 | 92 |
XM_005267044.2 | XP_005267101.1 | 1 - 2 | 275 | 92 | |
XM_011535925.2 | XP_011534227.1 | 1 - 2 | 275 | 92 | |
GenBank mRNA | BC030636.2 | - | 1 - 2 | 235 | 92 |
BM828521.1 | - | 1 - 2 | 248 | 92 | |
DA790125.1 | - | 1 - 2 | 275 | 92 |