The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001184937.1 | NP_001171866.1 | 12 - 13 | 1231 | 142 |
NM_001184938.2 | NP_001171867.1 | 12 - 13 | 1521 | 142 | |
NM_001184939.1 | NP_001171868.1 | 12 - 13 | 1231 | 142 | |
NM_020909.3 | NP_065960.2 | 12 - 13 | 1231 | 142 | |
NR_135920.1 | 12 - 13 | 1231 | 142 | ||
XM_006712651.1 | XP_006712714.1 | 12 - 13 | 1211 | 142 | |
XM_006712652.1 | XP_006712715.1 | 12 - 13 | 1526 | 142 | |
XM_006712653.1 | XP_006712716.1 | 12 - 13 | 1231 | 142 | |
XM_011511529.2 | XP_011509831.1 | 12 - 13 | 1211 | 142 | |
XM_017004567.1 | XP_016860056.1 | 12 - 13 | 1067 | 142 | |
XM_017004568.1 | XP_016860057.1 | 12 - 13 | 1231 | 142 | |
GenBank mRNA | AB046768.1 | - | 5 - 6 | 480 | 142 |
AK023019.1 | - | 12 - 13 | 1182 | 142 | |
AK290895.1 | - | 12 - 13 | 1141 | 142 | |
BC032822.2 | - | 12 - 13 | 1521 | 142 | |
BC054508.1 | - | 12 - 13 | 1166 | 142 |