Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
EPB41L5
Entrez Gene ID
57669
Gene Name
erythrocyte membrane protein band 4.1 like 5
Gene Aliases
BE37, YMO1, YRT
Chromosome Location
Chr.2: 120013028 - 120179121 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001184937.1 NP_001171866.1 20 - 21 1978 153
NM_020909.3 NP_065960.2 20 - 21 1978 153
XM_006712651.1 XP_006712714.1 20 - 21 1958 153
XM_006712652.1 XP_006712715.1 20 - 21 2273 153
XM_006712653.1 XP_006712716.1 20 - 21 1978 153
XM_011511529.2 XP_011509831.1 20 - 21 1958 153
XM_017004567.1 XP_016860056.1 20 - 21 1814 153
GenBank mRNA AB046768.1 - 13 - 14 1227 153
AK023019.1 - 20 - 21 1929 153
AK304831.1 - 3 - 4 256 153
AK307458.1 - 2 - 3 199 153
BC054508.1 - 20 - 21 1913 153

View Full Product Details