The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001278529.1 | NP_001265458.1 | 12 - 13 | 2665 | 112 |
NM_001278530.1 | NP_001265459.1 | 12 - 13 | 2471 | 112 | |
NM_018071.4 | NP_060541.3 | 12 - 13 | 2638 | 112 | |
XM_005267844.2 | XP_005267901.1 | 12 - 13 | 2674 | 112 | |
XM_011536937.2 | XP_011535239.1 | 12 - 13 | 2674 | 112 | |
XM_011536938.2 | XP_011535240.1 | 12 - 13 | 2676 | 112 | |
XM_017021434.1 | XP_016876923.1 | 13 - 14 | 2931 | 112 | |
XM_017021435.1 | XP_016876924.1 | 11 - 12 | 3148 | 112 | |
XM_017021436.1 | XP_016876925.1 | 8 - 9 | 931 | 112 | |
XM_017021437.1 | XP_016876926.1 | 8 - 9 | 931 | 112 | |
GenBank mRNA | AK024463.1 | - | 10 - 11 | 1876 | 112 |
AK074057.1 | - | 12 - 13 | 2583 | 112 | |
AK126755.1 | - | 8 - 9 | 733 | 112 | |
BC142692.1 | - | 12 - 13 | 2608 | 112 | |
BC151212.1 | - | 12 - 13 | 2608 | 112 |