The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001278529.1 | NP_001265458.1 | 1 - 2 | 138 | 145 |
NM_001278530.1 | NP_001265459.1 | 1 - 2 | 138 | 145 | |
NM_018071.4 | NP_060541.3 | 1 - 2 | 138 | 145 | |
XM_005267844.2 | XP_005267901.1 | 1 - 2 | 174 | 145 | |
XM_011536937.2 | XP_011535239.1 | 1 - 2 | 174 | 145 | |
XM_011536938.2 | XP_011535240.1 | 1 - 2 | 176 | 145 | |
XM_017021435.1 | XP_016876924.1 | 1 - 2 | 175 | 145 | |
GenBank mRNA | BX248768.1 | - | 1 - 2 | 125 | 145 |
HY010070.1 | - | 1 - 2 | 139 | 145 |