The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001303134.1 | NP_001290063.1 | 6 - 7 | 1192 | 167 |
NM_022494.2 | NP_071939.1 | 6 - 7 | 1204 | 167 | |
XM_005270051.4 | XP_005270108.1 | 6 - 7 | 2139 | 167 | |
XM_006717948.3 | XP_006718011.1 | 6 - 7 | 1310 | 167 | |
XM_011540091.2 | XP_011538393.1 | 6 - 7 | 1192 | 167 | |
XM_011540092.2 | XP_011538394.1 | 6 - 7 | 1109 | 167 | |
XM_011540093.1 | XP_011538395.1 | 6 - 7 | 1218 | 167 | |
XM_011540094.2 | XP_011538396.1 | 6 - 7 | 1077 | 167 | |
XM_017016561.1 | XP_016872050.1 | 6 - 7 | 1130 | 167 | |
XM_017016562.1 | XP_016872051.1 | 6 - 7 | 1298 | 167 | |
XM_017016563.1 | XP_016872052.1 | 6 - 7 | 1111 | 167 | |
XM_017016564.1 | XP_016872053.1 | 6 - 7 | 1310 | 167 | |
XM_017016565.1 | XP_016872054.1 | 6 - 7 | 1376 | 167 | |
XM_017016566.1 | XP_016872055.1 | 6 - 7 | 1189 | 167 | |
XM_017016568.1 | XP_016872057.1 | 6 - 7 | 1015 | 167 | |
GenBank mRNA | AF267740.1 | - | 6 - 7 | 1158 | 167 |
AK025605.1 | - | 6 - 7 | 1159 | 167 | |
AK223086.1 | - | 6 - 7 | 1158 | 167 | |
AK308026.1 | - | 6 - 7 | 1352 | 167 | |
BC007213.1 | - | 6 - 7 | 1134 | 167 | |
BC017434.2 | - | 6 - 7 | 1134 | 167 | |
BX647270.1 | - | 5 - 6 | 2632 | 167 |