The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | XM_006722885.3 | XP_006722948.2 | 1 - 2 | 409 | 76 |
XM_011528286.2 | XP_011526588.1 | 1 - 2 | 373 | 76 | |
XM_017027300.1 | XP_016882789.1 | 1 - 2 | 371 | 76 | |
XM_017027301.1 | XP_016882790.1 | 1 - 2 | 235 | 76 |