The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_145798.2 | NP_665741.1 | 20 - 21 | 2378 | 64 |
XM_017024122.1 | XP_016879611.1 | 20 - 21 | 2400 | 64 | |
XM_017024123.1 | XP_016879612.1 | 20 - 21 | 2518 | 64 | |
XM_017024124.1 | XP_016879613.1 | 19 - 20 | 2365 | 64 | |
XM_017024125.1 | XP_016879614.1 | 10 - 11 | 1301 | 64 | |
GenBank mRNA | AB208886.1 | - | 8 - 9 | 3225 | 64 |
AF323729.1 | - | 20 - 21 | 2367 | 64 | |
AF392446.1 | - | 20 - 21 | 2431 | 64 | |
BC028010.1 | - | 12 - 13 | 1566 | 64 | |
BC065482.1 | - | 20 - 21 | 2350 | 64 |