Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SPTBN1
Entrez Gene ID
6711
Gene Name
spectrin beta, non-erythrocytic 1
Gene Aliases
ELF, HEL102, SPTB2, betaSpII
Chromosome Location
Chr.2: 54456317 - 54671446 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_003128.2 NP_003119.2 15 - 16 3056 61
NM_178313.2 NP_842565.2 14 - 15 3153 61
XM_005264517.2 XP_005264574.1 15 - 16 2965 61
XM_005264518.1 XP_005264575.1 14 - 15 3811 61
XM_006712087.2 XP_006712150.1 15 - 16 2929 61
XM_017004779.1 XP_016860268.1 15 - 16 3072 61
XM_017004780.1 XP_016860269.1 15 - 16 3026 61
XM_017004781.1 XP_016860270.1 15 - 16 3027 61
GenBank mRNA AB209748.1 - 15 - 16 2934 61
AB371586.1 - 15 - 16 3046 61
AF327441.1 - 14 - 15 3153 61
BC137282.1 - 14 - 15 3063 61
BC137283.1 - 14 - 15 3063 61
EU794662.1 - 15 - 16 3056 61
S65762.1 - 7 - 8 1931 61

View Full Product Details