The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003128.2 | NP_003119.2 | 21 - 22 | 4743 | 87 |
NM_178313.2 | NP_842565.2 | 20 - 21 | 4840 | 87 | |
XM_005264517.2 | XP_005264574.1 | 21 - 22 | 4652 | 87 | |
XM_005264518.1 | XP_005264575.1 | 20 - 21 | 5498 | 87 | |
XM_006712087.2 | XP_006712150.1 | 21 - 22 | 4616 | 87 | |
XM_017004779.1 | XP_016860268.1 | 21 - 22 | 4759 | 87 | |
XM_017004780.1 | XP_016860269.1 | 21 - 22 | 4713 | 87 | |
XM_017004781.1 | XP_016860270.1 | 21 - 22 | 4714 | 87 | |
GenBank mRNA | AB209748.1 | - | 21 - 22 | 4621 | 87 |
AB371586.1 | - | 21 - 22 | 4733 | 87 | |
AF327441.1 | - | 20 - 21 | 4840 | 87 | |
BC137282.1 | - | 20 - 21 | 4750 | 87 | |
BC137283.1 | - | 20 - 21 | 4750 | 87 | |
EU794662.1 | - | 21 - 22 | 4743 | 87 | |
JQ599728.1 | - | 20 - 21 | 4494 | 87 | |
S65762.1 | - | 13 - 14 | 3618 | 87 |