The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003128.2 | NP_003119.2 | 10 - 11 | 1428 | 100 |
NM_178313.2 | NP_842565.2 | 9 - 10 | 1525 | 100 | |
XM_005264517.2 | XP_005264574.1 | 10 - 11 | 1337 | 100 | |
XM_005264518.1 | XP_005264575.1 | 9 - 10 | 2183 | 100 | |
XM_006712087.2 | XP_006712150.1 | 10 - 11 | 1301 | 100 | |
XM_017004779.1 | XP_016860268.1 | 10 - 11 | 1444 | 100 | |
XM_017004780.1 | XP_016860269.1 | 10 - 11 | 1398 | 100 | |
XM_017004781.1 | XP_016860270.1 | 10 - 11 | 1399 | 100 | |
GenBank mRNA | AB209748.1 | - | 10 - 11 | 1306 | 100 |
AB371586.1 | - | 10 - 11 | 1418 | 100 | |
AF327441.1 | - | 9 - 10 | 1525 | 100 | |
BC032061.1 | - | 10 - 11 | 1273 | 100 | |
BC137282.1 | - | 9 - 10 | 1435 | 100 | |
BC137283.1 | - | 9 - 10 | 1435 | 100 | |
EU794662.1 | - | 10 - 11 | 1428 | 100 | |
JQ599728.1 | - | 9 - 10 | 1179 | 100 | |
S65762.1 | - | 2 - 3 | 303 | 100 |