The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001128930.2 | NP_001122402.1 | 12 - 13 | 1877 | 74 |
NM_004901.4 | NP_004892.1 | 12 - 13 | 1901 | 74 | |
GenBank mRNA | AB002390.1 | - | 10 - 11 | 1431 | 74 |
AF016032.1 | - | 12 - 13 | 1806 | 74 | |
AJ131358.1 | - | 12 - 13 | 74 | ||
CN313713.1 | - | 1 - 2 | 117 | 74 |