The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001128930.2 | NP_001122402.1 | 5 - 6 | 835 | 65 |
NM_004901.4 | NP_004892.1 | 5 - 6 | 835 | 65 | |
GenBank mRNA | AB002390.1 | - | 3 - 4 | 389 | 65 |
AF016032.1 | - | 5 - 6 | 764 | 65 | |
AJ131358.1 | - | 5 - 6 | 732 | 65 | |
AK300464.1 | - | 5 - 6 | 743 | 65 | |
BC034477.2 | - | 5 - 6 | 799 | 65 | |
CN313716.1 | - | 3 - 4 | 391 | 65 |