The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001030272.2 | NP_001025443.1 | 15 - 16 | 1814 | 79 |
NM_001030273.2 | NP_001025444.1 | 16 - 17 | 1885 | 79 | |
NM_001178.5 | NP_001169.3 | 16 - 17 | 1869 | 79 | |
NM_001297719.1 | NP_001284648.1 | 16 - 17 | 1872 | 79 | |
NM_001297722.1 | NP_001284651.1 | 15 - 16 | 1817 | 79 | |
NM_001297724.1 | NP_001284653.1 | 16 - 17 | 1881 | 79 | |
XM_011520107.2 | XP_011518409.2 | 14 - 15 | 1915 | 79 | |
XM_011520112.2 | XP_011518414.1 | 17 - 18 | 1880 | 79 | |
XM_011520113.1 | XP_011518415.1 | 17 - 18 | 1877 | 79 | |
XM_017017739.1 | XP_016873228.1 | 14 - 15 | 1917 | 79 | |
XM_017017742.1 | XP_016873231.1 | 14 - 15 | 1620 | 79 | |
XM_017017743.1 | XP_016873232.1 | 14 - 15 | 1617 | 79 | |
XM_017017746.1 | XP_016873235.1 | 16 - 17 | 1835 | 79 | |
XM_017017747.1 | XP_016873236.1 | 15 - 16 | 1695 | 79 | |
XM_017017748.1 | XP_016873237.1 | 15 - 16 | 1749 | 79 | |
GenBank mRNA | AB000812.1 | - | 13 - 14 | 1453 | 79 |
AB000814.1 | - | 2 - 3 | 269 | 79 | |
AF044288.1 | - | 16 - 17 | 1782 | 79 | |
AK095749.1 | - | 16 - 17 | 1770 | 79 | |
AK128317.1 | - | 2 - 3 | 172 | 79 | |
AK291510.1 | - | 15 - 16 | 1730 | 79 | |
AK315174.1 | - | 16 - 17 | 1810 | 79 | |
BC016674.1 | - | 13 - 14 | 1416 | 79 | |
BC031214.1 | - | 16 - 17 | 1713 | 79 | |
BC041129.1 | - | 16 - 17 | 1793 | 79 | |
D89722.1 | - | 13 - 14 | 1429 | 79 | |
U51627.1 | - | 10 - 11 | 1411 | 79 | |
U60415.1 | - | 15 - 16 | 1716 | 79 |