Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PTH
Entrez Gene ID
5741
Gene Name
parathyroid hormone
Gene Aliases
PTH1
Chromosome Location
Chr.11: 13492045 - 13496175 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001316352.1 NP_001303281.1 1 - 2 133 84
GenBank mRNA BG188276.1 - 1 - 2 143 84

View Full Product Details