The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_019044.4 | NP_061917.3 | 17 - 18 | 1518 | 99 |
XM_011511359.1 | XP_011509661.1 | 17 - 18 | 1515 | 99 | |
XM_011511361.1 | XP_011509663.1 | 16 - 17 | 1272 | 99 | |
XM_011511362.1 | XP_011509664.1 | 16 - 17 | 1386 | 99 | |
GenBank mRNA | AB209993.1 | - | 17 - 18 | 1437 | 99 |
AK001858.1 | - | 5 - 6 | 312 | 99 | |
AK025611.1 | - | 16 - 17 | 1286 | 99 | |
AK057926.1 | - | 17 - 18 | 1379 | 99 | |
AK290722.1 | - | 17 - 18 | 1425 | 99 | |
AK309040.1 | - | 17 - 18 | 1425 | 99 | |
BC028609.1 | - | 17 - 18 | 1491 | 99 | |
BC093018.1 | - | 17 - 18 | 1408 | 99 |