The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_019044.4 | NP_061917.3 | 21 - 22 | 1807 | 155 |
XM_011511359.1 | XP_011509661.1 | 21 - 22 | 1804 | 155 | |
XM_011511361.1 | XP_011509663.1 | 20 - 21 | 1561 | 155 | |
XM_011511362.1 | XP_011509664.1 | 20 - 21 | 1675 | 155 | |
GenBank mRNA | AB209993.1 | - | 21 - 22 | 1726 | 155 |
AK001858.1 | - | 9 - 10 | 601 | 155 | |
AK025611.1 | - | 20 - 21 | 1575 | 155 | |
AK057926.1 | - | 21 - 22 | 1668 | 155 | |
AK290722.1 | - | 21 - 22 | 1714 | 155 | |
BC028609.1 | - | 21 - 22 | 1780 | 155 | |
BC093018.1 | - | 21 - 22 | 1697 | 155 |