The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_019044.4 | NP_061917.3 | 22 - 23 | 1890 | 96 |
XM_011511359.1 | XP_011509661.1 | 22 - 23 | 1887 | 96 | |
XM_011511361.1 | XP_011509663.1 | 21 - 22 | 1644 | 96 | |
XM_011511362.1 | XP_011509664.1 | 21 - 22 | 1758 | 96 | |
GenBank mRNA | AB209993.1 | - | 22 - 23 | 1809 | 96 |
AK001858.1 | - | 10 - 11 | 684 | 96 | |
AK025611.1 | - | 21 - 22 | 1658 | 96 | |
AK290722.1 | - | 22 - 23 | 1797 | 96 | |
BC028609.1 | - | 22 - 23 | 1863 | 96 | |
BC093018.1 | - | 22 - 23 | 1780 | 96 |