Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CCDC93
Entrez Gene ID
54520
Gene Name
coiled-coil domain containing 93
Gene Aliases
-
Chromosome Location
Chr.2: 117915478 - 118014163 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_019044.4 NP_061917.3 5 - 6 633 79
XM_006712600.2 XP_006712663.1 5 - 6 633 79
XM_011511359.1 XP_011509661.1 5 - 6 633 79
XM_011511361.1 XP_011509663.1 4 - 5 387 79
XM_011511362.1 XP_011509664.1 4 - 5 501 79
XM_011511363.2 XP_011509665.1 5 - 6 633 79
GenBank mRNA AB209993.1 - 5 - 6 552 79
AK025611.1 - 4 - 5 401 79
AK057926.1 - 5 - 6 494 79
AK290722.1 - 5 - 6 540 79
AK309040.1 - 5 - 6 540 79
BC028609.1 - 5 - 6 606 79
BC093018.1 - 5 - 6 523 79

View Full Product Details