Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CCDC93
Entrez Gene ID
54520
Gene Name
coiled-coil domain containing 93
Gene Aliases
-
Chromosome Location
Chr.2: 117915478 - 118014163 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_019044.4 NP_061917.3 9 - 10 920 73
XM_006712600.2 XP_006712663.1 9 - 10 920 73
XM_011511359.1 XP_011509661.1 9 - 10 920 73
XM_011511361.1 XP_011509663.1 8 - 9 674 73
XM_011511362.1 XP_011509664.1 8 - 9 788 73
XM_011511363.2 XP_011509665.1 9 - 10 920 73
GenBank mRNA AB209993.1 - 9 - 10 839 73
AK025611.1 - 8 - 9 688 73
AK057926.1 - 9 - 10 781 73
AK290722.1 - 9 - 10 827 73
AK309040.1 - 9 - 10 827 73
BC028609.1 - 9 - 10 893 73
BC093018.1 - 9 - 10 810 73

View Full Product Details